siRNA Details
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013791.2 | NP_038819.1 | 10 | 1125 | ||
GenBank mRNA | AK077541.1 | - | 10 | 1120 | ||
AK136123.1 | - | 7 | 810 | |||
BC013703.1 | - | 10 | 1109 | 3668925 | ||
U72194.1 | - | 10 | 1108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
