siRNA Details
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013791.2 | NP_038819.1 | 6 | 653 | ||
GenBank mRNA | AK037967.1 | - | 6 | 654 | ||
AK077541.1 | - | 6 | 649 | |||
AK136123.1 | - | 3 | 336 | |||
AK155457.1 | - | 6 | 634 | |||
AK164822.1 | - | 6 | 649 | |||
AK165239.1 | - | 6 | 653 | |||
BC013703.1 | - | 6 | 637 | 3668925 | ||
U72194.1 | - | 6 | 636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
