siRNA Details
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077595.2 | NP_001071063.1 | 9 | 5303 | ||
NM_001077596.2 | NP_001071064.1 | 9 | 5351 | |||
NM_015756.2 | NP_056571.2 | 10 | 5675 | |||
GenBank mRNA | AF199421.1 | - | 10 | 5693 | ||
AF199422.1 | - | 9 | 5261 | |||
AK003320.1 | - | 1 | 195 | |||
AK049347.1 | - | 6 | 3209 | |||
AK129371.1 | - | 6 | 4681 | |||
AK147493.1 | - | 9 | 5286 | |||
AK164918.1 | - | 9 | 5262 | |||
BC003909.1 | - | 4 | 947 | |||
BC145113.1 | - | 11 | 5092 | 9053661 |
