siRNA Details
Gene Symbol: Ubl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145405.2 | NP_663380.1 | 2, 3 | 250 | ||
GenBank mRNA | AJ627038.1 | - | 2, 3 | 211 | ||
AK048689.1 | - | 1, 2 | 87 | |||
AK146110.1 | - | 240 | ||||
BC010817.1 | - | 2, 3 | 229 | 4215699 | ||
BM932729.1 | - | 2, 3 | 224 | |||
BY107969.1 | - | 1, 2 | 115 | |||
BY273712.1 | - | 1, 2 | 145 |
Gene Symbol: Gm38419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278271.1 | NP_001265200.1 | 5, 6 | 849 | ||
NM_001278272.1 | NP_001265201.1 | 5, 6 | 813 | |||
GenBank mRNA | EU599043.1 | - | 5, 6 | 718 | ||
EU599044.1 | - | 5, 6 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
