siRNA Details
Gene Symbol: Snf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033568.2 | NP_291046.1 | 6, 7 | 667 | ||
GenBank mRNA | AK013058.1 | - | 6, 7 | 669 | ||
AK020203.1 | - | 4, 5 | 357 | |||
BC003938.1 | - | 6, 7 | 639 | 3486703 | ||
BC030490.1 | - | 1, 2 | 632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
