siRNA Details
Gene Symbol: Spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138584.2 | NP_613050.1 | 3, 4 | 473 | ||
GenBank mRNA | AK009332.1 | - | 3, 4 | 465 | ||
AK011985.1 | - | 3, 4 | 440 | |||
AK090017.1 | - | 3, 4 | 461 | |||
AK149930.1 | - | 3, 4 | 447 | |||
AK150215.1 | - | 3, 4 | 474 | |||
AK150581.1 | - | 3, 4 | 451 | |||
AK153284.1 | - | 3, 4 | 435 | |||
AK170666.1 | - | 3, 4 | 466 | |||
AK179732.1 | - | 18 | ||||
AK199934.1 | - | 18 | ||||
BC099434.1 | - | 2, 3 | 238 | 30714184 | ||
BC132377.1 | - | 3, 4 | 450 | 40130654 | ||
BC132403.1 | - | 3, 4 | 450 | 40130680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
