siRNA Details
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB041555.1 | - | 5 | 1028 | ||
AK003799.1 | - | 3 | 1599 | |||
AK028145.1 | - | 3 | 1601 | |||
AK029529.1 | - | 3 | 1597 | |||
AK160993.1 | - | 3 | 1599 | |||
AK169598.1 | - | 5 | 1052 | |||
AK169760.1 | - | 3 | 1599 | |||
BC043057.1 | - | 3 | 1583 | 6419597 | ||
BC046231.1 | - | 3 | 1500 | 6813388 | ||
BC054728.1 | - | 3 | 1571 | 6419723 | ||
BC056986.1 | - | 3 | 1532 | 6816649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
