siRNA Details
Gene Symbol: Fam3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138587.4 | NP_613053.3 | 3 | 193 | ||
GenBank mRNA | AK154498.1 | - | 3 | 223 | ||
AK170887.1 | - | 3 | 242 | |||
AY424275.1 | - | 3 | 193 | |||
BC009086.1 | - | 3 | 193 | 2811008 | ||
BC055853.1 | - | 4 | 431 | 4012164 | ||
BC069880.1 | - | 2 | 319 |

More Information
Other Species Targeted

Gene Ontology Categories:
