siRNA Details
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133345.2 | NP_579923.1 | 6 | 601 | ||
GenBank mRNA | AK002821.1 | - | 6 | 603 | ||
AK004347.1 | - | 601 | ||||
AK009267.1 | - | 6 | 602 | |||
AK031633.1 | - | 6 | 609 | |||
AK050522.1 | - | 6 | 614 | |||
AY035880.1 | - | 5 | 793 | |||
AY035881.1 | - | 6 | 588 | |||
BC009127.1 | - | 6 | 589 | 3708722 | ||
BC082781.1 | - | 2 | 2786 |

More Information
Other Species Targeted

Gene Ontology Categories:
