siRNA Details
Gene Symbol: Mrpl50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178603.4 | NP_848718.1 | 2 | 277 | ||
GenBank mRNA | AK002646.1 | - | 2 | 265 | ||
AK002686.1 | - | 2 | 279 | |||
AK007405.1 | - | 2 | 274 | |||
AK031229.1 | - | 2 | 278 | |||
AK049421.1 | - | 2 | 267 | |||
AK075787.1 | - | 2 | 259 | |||
BC021318.1 | - | 2 | 288 | 5035369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
