siRNA Details
Gene Symbol: D6Wsu163e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138594.3 | NP_613060.1 | 9, 10 | 1244 | ||
GenBank mRNA | AK019434.1 | - | 1, 2 | 134 | ||
AK037095.1 | - | 9, 10 | 1249 | |||
AK045541.1 | - | 9, 10 | 1245 | |||
AK046707.1 | - | 9, 10 | 1232 | |||
AK137201.1 | - | 9, 10 | 1235 | |||
BC016429.1 | - | 8, 9 | 1192 | 4486831 |
