siRNA Details
Gene Symbol: Ept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027652.2 | NP_081928.2 | 2 | 229 | ||
GenBank mRNA | AK014530.1 | - | 2 | 175 | ||
AK040494.1 | - | 2 | 208 | |||
AK040745.1 | - | 2 | 138 | |||
AK048011.1 | - | 2 | 178 | |||
AK079325.1 | - | 2 | 205 | |||
AK083973.1 | - | 2 | 205 | |||
AK122544.1 | - | 2 | 145 | |||
AK140416.1 | - | 2 | 201 | |||
BC106097.1 | - | 2 | 198 | 6485810 | ||
BC115783.1 | - | 2 | 168 | 40104916 | ||
BC117551.1 | - | 2 | 163 | 40104922 | ||
BY136287.1 | - | 2 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
