siRNA Details
Gene Symbol: Med10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138596.2 | NP_613062.1 | 3 | 314 | ||
GenBank mRNA | AK003862.1 | - | 3 | 308 | ||
AK013992.1 | - | 3 | 318 | |||
AK156498.1 | - | 3 | 323 | |||
AK215905.1 | - | 3 | 162 | |||
BC048638.1 | - | 3 | 307 | 6771841 | ||
BC056438.1 | - | 3 | 303 | 6818638 | ||
BC059231.1 | - | 2 | 2796 | |||
BC089312.1 | - | 2 | 1859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
