siRNA Details
Gene Symbol: Mydgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080837.2 | NP_543027.1 | 1, 2 | 182 | ||
GenBank mRNA | AK004074.1 | - | 1, 2 | 182 | ||
AK008289.1 | - | 1, 2 | 168 | |||
AK008854.1 | - | 1, 2 | 163 | |||
AK018016.1 | - | 1, 2 | 165 | |||
AK133947.1 | - | 166 | ||||
AK148083.1 | - | 1, 2 | 182 | |||
AK162834.1 | - | 1, 2 | 170 | |||
AY038184.1 | - | 1, 2 | 178 | |||
BC132127.1 | - | 1, 2 | 203 | 40130404 | ||
BC132129.1 | - | 1, 2 | 203 | 40130406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
