siRNA Details
Gene Symbol: Apon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133996.3 | NP_598757.1 | 2 | 865 | ||
GenBank mRNA | AK083289.1 | - | 1 | 1001 | ||
BC022137.1 | - | 1 | 932 | |||
BC030734.1 | - | 1 | 805 | |||
BC034832.1 | - | 1 | 964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
