siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 3 | 122 | ||
GenBank mRNA | AB027568.1 | - | 3 | 123 | ||
AK034521.1 | - | 132 | ||||
AK035044.1 | - | 3 | 123 | |||
AK039131.1 | - | 3 | 167 | |||
AK052225.1 | - | 3 | 113 | |||
AK087844.1 | - | 3 | 226 | |||
AK136486.1 | - | 3 | 123 | |||
BC015246.1 | - | 3 | 139 | 4221866 | ||
BC023354.1 | - | 3 | 98 | 5253545 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
