siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 3 | 119 | ||
GenBank mRNA | AB027568.1 | - | 3 | 120 | ||
AK034521.1 | - | 129 | ||||
AK035044.1 | - | 3 | 120 | |||
AK039131.1 | - | 3 | 164 | |||
AK052225.1 | - | 3 | 110 | |||
AK087844.1 | - | 3 | 223 | |||
AK136486.1 | - | 3 | 120 | |||
BC015246.1 | - | 3 | 136 | 4221866 | ||
BC023354.1 | - | 3 | 95 | 5253545 |
