siRNA Details
Gene Symbol: Tpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013861.3 | NP_038889.1 | 3 | 141 | ||
GenBank mRNA | AB027568.1 | - | 3 | 142 | ||
AK028431.1 | - | 3 | 164 | |||
AK034521.1 | - | 151 | ||||
AK035044.1 | - | 3 | 142 | |||
AK039131.1 | - | 3 | 186 | |||
AK052225.1 | - | 3 | 132 | |||
AK087844.1 | - | 3 | 245 | |||
AK136486.1 | - | 3 | 142 | |||
BC015246.1 | - | 3 | 158 | 4221866 | ||
BC023354.1 | - | 3 | 117 | 5253545 |
