siRNA Details
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164274.1 | NP_001157746.1 | 3 | 1439 | ||
NM_013720.2 | NP_038748.2 | 3 | 1439 | |||
GenBank mRNA | AF205935.1 | - | 2 | 1253 | ||
AK032570.1 | - | 3 | 1420 | |||
AK046252.1 | - | 3 | 1440 | |||
AK132911.1 | - | 1 | 1716 | |||
AK133775.1 | - | 3 | 1656 | |||
AK139919.1 | - | 3 | 1391 | |||
AK145491.1 | - | 3 | 1388 | |||
AK162302.1 | - | 3 | 1436 | |||
AK172962.1 | - | 3 | 1362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
