siRNA Details
Gene Symbol: Rabgap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038621.2 | NP_001033710.1 | 2 | 454 | ||
NM_013862.5 | NP_038890.3 | 19 | 2559 | |||
GenBank mRNA | AK035796.1 | - | 19 | 2560 | ||
AK147634.1 | - | 2 | 455 | |||
BC145811.1 | - | 19 | 2404 | 40131114 | ||
BC145813.1 | - | 19 | 2404 | 40131116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
