siRNA Details
Gene Symbol: Hip1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145070.3 | NP_659507.3 | 5 | 562 | ||
GenBank mRNA | AF221713.1 | - | 5 | 427 | ||
AK014542.1 | - | 5 | 540 | |||
AK129182.1 | - | 5 | 518 | |||
AK146663.1 | - | 5 | 443 | |||
BC062914.1 | - | 5 | 443 | |||
BC065101.1 | - | 5 | 551 | 30535741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
