siRNA Details
Gene Symbol: Sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013873.3 | NP_038901.3 | 7 | 946 | ||
GenBank mRNA | AF059257.1 | - | 7 | 822 | ||
AK003034.1 | - | 7 | 807 | |||
AK010293.1 | - | 8 | 931 | |||
AK032195.1 | - | 7 | 832 | |||
AK046368.1 | - | 7 | 825 | |||
AK159034.1 | - | 7 | 838 | |||
BC051132.1 | - | 7 | 819 | 6531493 | ||
BC054757.1 | - | 7 | 946 | 6413084 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
