siRNA Details
Gene Symbol: Cabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160252.1 | NP_001153724.1 | 5 | 490 | ||
NM_001160253.1 | NP_001153725.1 | 5 | 772 | |||
NM_013878.2 | NP_038906.2 | 6 | 649 | |||
GenBank mRNA | AF169156.1 | - | 5 | 518 | ||
AF169157.1 | - | 6 | 677 | |||
AK007488.1 | - | 5 | 772 | |||
BC107029.2 | - | 6 | 649 | 40050356 |

More Information
Other Species Targeted

Gene Ontology Categories:
