siRNA Details
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013887.2 | NP_038915.1 | 9 | 1559 | ||
GenBank mRNA | AF147789.1 | - | 9 | 1559 | ||
BC139827.1 | - | 9 | 1500 | 40142922 | ||
EU303118.1 | - | 9 | 1375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
