siRNA Details
Gene Symbol: Scd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024450.2 | NP_077770.1 | 6 | 1520 | ||
GenBank mRNA | AF272037.1 | - | 6 | 1509 | ||
AK078675.1 | - | 6 | 1543 | |||
AK078713.1 | - | 6 | 1541 | |||
AK136277.1 | - | 6 | 1537 | |||
BC118033.1 | - | 6 | 1418 | 40106893 | ||
BC118045.1 | - | 6 | 1418 | 40106896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
