siRNA Details
Gene Symbol: Scd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024450.2 | NP_077770.1 | 6 | 1859 | ||
GenBank mRNA | AF272037.1 | - | 6 | 1846 | ||
AK078675.1 | - | 6 | 1882 | |||
AK136277.1 | - | 6 | 1876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
