siRNA Details
Gene Symbol: Rnf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033043.1 | NP_001028215.1 | 7 | 659 | ||
GenBank mRNA | AF190166.1 | - | 7 | 790 | ||
AF285585.1 | - | 7 | 659 | |||
AK136196.1 | - | 7 | 657 | |||
AY854010.1 | - | 7 | 659 | |||
AY854011.1 | - | 7 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
