siRNA Details
Gene Symbol: Timm8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013897.2 | NP_038925.1 | 2 | 163 | ||
GenBank mRNA | AA204614.1 | - | 2 | 152 | ||
AF196314.1 | - | 152 | ||||
AK003382.1 | - | 2 | 122 | |||
AK004190.1 | - | 152 | ||||
AK012925.1 | - | 2 | 130 | |||
AK018718.1 | - | 2 | 130 | |||
AK019422.1 | - | 2 | 138 | |||
AK169258.1 | - | 2 | 142 | |||
BC029239.1 | - | 2 | 116 | 4237232 | ||
BY100901.1 | - | 2 | 167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
