siRNA Details
Gene Symbol: Zfp330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145600.1 | NP_663575.1 | 10 | 962 | ||
GenBank mRNA | AK029489.1 | - | 9 | 787 | ||
AK037748.1 | - | 4 | 2893 | |||
AK050534.1 | - | 10 | 972 | |||
BC008086.1 | - | 10 | 962 | 3586161 | ||
BC030675.1 | - | 10 | 960 | 1068780 | ||
BC037025.1 | - | 6 | 2319 | |||
BC083081.1 | - | 10 | 996 | 6510396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
