siRNA Details
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 5 | 887 | ||
GenBank mRNA | AJ299404.1 | - | 5 | 855 | ||
AK007814.1 | - | 5 | 876 | |||
AK009693.1 | - | 5 | 905 | |||
AK017908.1 | - | 5 | 832 | |||
AK151048.1 | - | 5 | 873 | |||
AK151390.1 | - | 6 | 924 | |||
AK152243.1 | - | 5 | 852 | |||
AK152358.1 | - | 5 | 888 | |||
AK152484.1 | - | 5 | 888 | |||
AK153391.1 | - | 5 | 873 | |||
AK170296.1 | - | 5 | 882 | |||
AK172355.1 | - | 5 | 862 | |||
BC025525.1 | - | 5 | 871 | |||
BC052851.1 | - | 5 | 1176 | |||
BC071216.1 | - | 5 | 868 | 30439199 |
