siRNA Details
Gene Symbol: Zfp354c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013922.4 | NP_038950.3 | 5 | 812 | ||
GenBank mRNA | AF192804.1 | - | 4 | 607 | ||
AK086760.1 | - | 5 | 775 | |||
AK145882.1 | - | 5 | 806 | |||
AK220194.1 | - | 5 | 780 | |||
BC079908.1 | - | 5 | 812 | 30654724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
