siRNA Details
Gene Symbol: Schip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013928.5 | NP_038956.2 | 1 | 267 | ||
GenBank mRNA | AK021361.1 | - | 1 | 128 | ||
BC060529.1 | - | 1 | 201 | |||
BY243167.1 | - | 1 | 268 | |||
KM233716.1 | - | 1 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
