siRNA Details
Gene Symbol: Vapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013933.3 | NP_038961.2 | 2 | 354 | ||
GenBank mRNA | AF115503.1 | - | 2 | 76 | ||
AF157497.1 | - | 2 | 295 | |||
AK003576.1 | - | 2 | 295 | |||
AK012357.1 | - | 2 | 109 | |||
AK088132.1 | - | 2 | 333 | |||
AK159582.1 | - | 2 | 296 | |||
AK167381.1 | - | 2 | 355 | |||
AK168824.1 | - | 2 | 301 | |||
AK183351.1 | - | 2 | 121 | |||
AK189268.1 | - | 83 | ||||
BC003866.1 | - | 2 | 340 | 3490082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
