siRNA Details
Gene Symbol: Nox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285833.1 | NP_001272762.1 | 20 | 1976 | ||
NM_001285835.1 | NP_001272764.1 | 18 | 1859 | |||
NM_015760.5 | NP_056575.1 | 18 | 1863 | |||
GenBank mRNA | AB041034.1 | - | 18 | 1714 | ||
AB042745.1 | - | 18 | 1714 | |||
AF218723.1 | - | 18 | 1714 | |||
AF261944.1 | - | 18 | 1718 | |||
AF276957.1 | - | 18 | 1725 | |||
AK050371.1 | - | 18 | 1718 | |||
AK149398.1 | - | 18 | 1713 | |||
AK149455.1 | - | 18 | 1714 | |||
AK149492.1 | - | 18 | 1714 | |||
BC021378.1 | - | 20 | 1824 | |||
BC138918.1 | - | 18 | 1710 | 8861940 | ||
BC145462.1 | - | 18 | 1706 | |||
EU861989.1 | - | 9 | 745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
