siRNA Details
Gene Symbol: Nox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285833.1 | NP_001272762.1 | 7 | 730 | ||
NM_001285835.1 | NP_001272764.1 | 7 | 730 | |||
NM_015760.5 | NP_056575.1 | 7 | 734 | |||
GenBank mRNA | AB041034.1 | - | 7 | 585 | ||
AB042745.1 | - | 7 | 585 | |||
AF218723.1 | - | 7 | 585 | |||
AF261944.1 | - | 7 | 589 | |||
AF276957.1 | - | 7 | 596 | |||
AK050371.1 | - | 7 | 589 | |||
AK052905.1 | - | 7 | 585 | |||
AK085509.1 | - | 7 | 590 | |||
AK149398.1 | - | 7 | 585 | |||
AK149455.1 | - | 7 | 585 | |||
AK149492.1 | - | 7 | 585 | |||
BC021378.1 | - | 7 | 578 | |||
BC138918.1 | - | 7 | 581 | 8861940 | ||
BC145462.1 | - | 7 | 577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
