siRNA Details
Gene Symbol: Txnrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042513.1 | NP_001035978.1 | 8, 9 | 1003 | ||
NM_001042514.1 | NP_001035979.1 | 7, 8 | 1091 | |||
NM_001042523.1 | NP_001035988.1 | 8, 9 | 1220 | |||
NM_015762.2 | NP_056577.2 | 7, 8 | 896 | |||
GenBank mRNA | AB027565.1 | - | 7, 8 | 923 | ||
AF333036.1 | - | 8, 9 | 1220 | |||
AK011902.1 | - | 8, 9 | 1004 | |||
AK149625.1 | - | 8, 9 | 1004 | |||
AK151927.1 | - | 8, 9 | 1004 | |||
AK168356.1 | - | 8, 9 | 1004 | |||
AK181192.1 | - | 1, 2 | 105 | |||
AK183038.1 | - | 1, 2 | 105 | |||
AK185436.1 | - | 1, 2 | 105 | |||
AK189898.1 | - | 1, 2 | 105 | |||
AK192872.1 | - | 1, 2 | 105 | |||
AK195172.1 | - | 1, 2 | 105 | |||
AK198860.1 | - | 1, 2 | 105 | |||
AK203979.1 | - | 1, 2 | 105 | |||
AK204938.1 | - | 1, 2 | 105 | |||
BC020274.1 | - | 7, 8 | 858 | |||
BC037643.1 | - | 7, 8 | 1176 | 5059831 |
