siRNA Details
Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015767.3 | NP_056582.1 | 5 | 695 | ||
GenBank mRNA | AF218416.1 | - | 5 | 683 | ||
AK004882.1 | - | 5 | 676 | |||
AK050374.1 | - | 5 | 696 | |||
BC026882.1 | - | 5 | 582 | |||
BC043705.1 | - | 1 | 250 | |||
BC099868.1 | - | 5 | 694 | 40040166 | ||
BC099869.1 | - | 5 | 696 | 40040167 | ||
BC099870.1 | - | 5 | 696 | 40040169 | ||
BC099871.1 | - | 5 | 695 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
