siRNA Details
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037539.2 | NP_001032628.1 | 2 | 219 | ||
NM_015768.2 | NP_056583.1 | 2 | 219 | |||
GenBank mRNA | AF182064.1 | - | 2 | 219 | ||
AF182066.1 | - | 2 | 219 | |||
AF487280.1 | - | 2 | 210 | |||
AK015462.1 | - | 2 | 181 | |||
BC116911.1 | - | 2 | 188 | 40126230 | ||
BC144863.1 | - | 2 | 200 | 9053411 | ||
BY232101.1 | - | 2 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
