siRNA Details
Gene Symbol: Sap30l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081168.1 | NP_001074637.1 | 1 | 636 | ||
GenBank mRNA | AK010234.1 | - | 1 | 124 | ||
BC051686.1 | - | 1 | 636 | 6433799 | ||
BC094930.1 | - | 2 | 355 | |||
BC117029.1 | - | 1 | 350 | 40126348 | ||
BC117031.1 | - | 1 | 350 | 40126350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
