siRNA Details
Gene Symbol: Fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015795.1 | NP_056610.1 | 6 | 805 | ||
GenBank mRNA | AF176531.1 | - | 6 | 805 | ||
AK043554.1 | - | 6 | 775 | |||
AK081947.1 | - | 6 | 914 | |||
AK164657.1 | - | 6 | 888 | |||
BC086690.1 | - | 6 | 769 | 30683387 | ||
BC115890.1 | - | 6 | 763 | 40103787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
