siRNA Details
Gene Symbol: Fbxo17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015796.2 | NP_056611.2 | 6 | 1104 | ||
GenBank mRNA | AF176532.1 | - | 6 | 1021 | ||
AK133408.1 | - | 6 | 1064 | |||
BC026470.1 | - | 6 | 1090 | 4486850 |

More Information
Other Species Targeted

Gene Ontology Categories:
