siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 20 | 2412 | ||
NM_015805.3 | NP_056620.2 | 20 | 2412 | |||
GenBank mRNA | AF155913.1 | - | 19 | 2140 | ||
AK030485.1 | - | 19 | 2072 | |||
AK035355.1 | - | 1 | 1604 | |||
BC003246.1 | - | 2 | 169 | |||
BC057000.1 | - | 19 | 2260 | |||
BC079626.1 | - | 20 | 2337 | 30536500 |
