siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 12, 13 | 1386 | ||
NM_015805.3 | NP_056620.2 | 12, 13 | 1386 | |||
GenBank mRNA | AF155913.1 | - | 11, 12 | 1114 | ||
AK030485.1 | - | 11, 12 | 1046 | |||
AK044331.1 | - | 11, 12 | 1284 | |||
BC057000.1 | - | 11, 12 | 1234 | |||
BC079626.1 | - | 12, 13 | 1311 | 30536500 |
