siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201569.1 | NP_001188498.1 | 7, 8 | 902 | ||
NM_015805.3 | NP_056620.2 | 7, 8 | 902 | |||
GenBank mRNA | AB206122.1 | - | 6, 7 | 630 | ||
AF155913.1 | - | 6, 7 | 630 | |||
AK030485.1 | - | 6, 7 | 563 | |||
AK044331.1 | - | 7, 8 | 877 | |||
BC057000.1 | - | 7, 8 | 827 | |||
BC079626.1 | - | 7, 8 | 827 | 30536500 |
