siRNA Details
Gene Symbol: Rgs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282061.1 | NP_001268990.1 | 15 | 1309 | ||
NM_015812.3 | NP_056627.1 | 15 | 1591 | |||
GenBank mRNA | AF061933.1 | - | 12 | 1036 | ||
AK052527.1 | - | 15 | 1592 | |||
AK053087.1 | - | 15 | 1310 | |||
BC038616.1 | - | 13 | 1136 | |||
BC125029.1 | - | 15 | 1295 | 40062355 | ||
BC125030.1 | - | 15 | 1295 | 40062356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
