siRNA Details
Gene Symbol: Ppap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005452.1 | - | 3 | 312 | ||
AK049581.1 | - | 3 | 355 | |||
AK177565.1 | - | 2 | 61 | |||
AK179023.1 | - | 2 | 116 | |||
AK208995.1 | - | 2 | 61 | |||
AK214880.1 | - | 2 | 85 | |||
BC010332.1 | - | 3 | 333 | 2654393 | ||
BC085104.1 | - | 3 | 271 | |||
BQ571983.1 | - | 2 | 158 | |||
CA540403.1 | - | 3 | 278 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF123611.1 | - | 347 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
