siRNA Details
Gene Symbol: Hs6st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015818.2 | NP_056633.2 | 2 | 1665 | ||
GenBank mRNA | AB024566.1 | - | 2 | 1656 | ||
AK144809.1 | - | 3 | 1585 | |||
AK157556.1 | - | 2 | 1529 | |||
AK165161.1 | - | 1 | 2705 | |||
AK165385.1 | - | 2 | 1068 | |||
AK166962.1 | - | 770 | ||||
AK167151.1 | - | 2 | 1094 | |||
BC052316.1 | - | 2 | 1665 | 6391204 |
