siRNA Details
Gene Symbol: Fbxl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015821.2 | NP_056636.2 | 3 | 644 | ||
GenBank mRNA | AF176523.1 | - | 3 | 569 | ||
AK053206.1 | - | 4 | 812 | |||
AK155194.1 | - | 3 | 605 | |||
AK170367.1 | - | 3 | 604 | |||
BC023902.1 | - | 2 | 509 | 5321302 | ||
CX222518.1 | - | 3 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
