siRNA Details
Gene Symbol: Hnrnpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016690.4 | NP_057899.2 | 6 | 1109 | ||
GenBank mRNA | AB017020.1 | - | 6 | 815 | ||
AK011767.1 | - | 6 | 799 | |||
AK172629.1 | - | 6 | 768 | |||
BC021374.1 | - | 6 | 767 | 5039077 | ||
CX219512.1 | - | 5 | 484 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
