siRNA Details
Gene Symbol: Uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016723.2 | NP_057932.2 | 3 | 206 | ||
GenBank mRNA | AB033370.1 | - | 3 | 179 | ||
AF247358.1 | - | 116 | ||||
AK050278.1 | - | 3 | 206 | |||
BC029242.1 | - | 3 | 148 | |||
BC048481.1 | - | 3 | 163 | 6773555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
